Learn More
Medchemexpress LLC β-nicotinamide adenine dinucleotide-d4 | 98.3% | 667.45 g/mol | C21H23D4N7O14P2 | 500 UG
SDP
Catalog No. 5000284053
Encompass_Preferred
Deuterium-labeled nicotinamide adenine dinucleotide (NAD+) supplied as a white to off-white solid for use as a tracer and internal standard in quantitative analytical methods such as NMR, GC-MS, and LC-MS. Supplied as a 500 μg powder and intended for research use; follow storage and handling instructions.
Catalog No. 50-002-84053 Supplier Medchemexpress LLC Supplier No. HYW654288500UG
Deuterium-labeled nicotinamide adenine dinucleotide (NAD+) supplied as a white to off-white solid for use as a tracer and internal standard in quantitative analytical methods such as NMR, GC-MS, and LC-MS. Supplied as a 500 μg powder and intended for research use; follow storage and handling instructions.