Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

β-Nicotinamide Adenine Dinucleotide , MP Biomedicals, LLC

Catalog No. MP210031901 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210031901 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210031901 Supplier MP Biomedicals, Inc Supplier No. 0210031901
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

β-Nicotinamide Adenine Dinucleotide is a hydrogen acceptor utilized for molecular biology and has a purity >93%.

  • Biologically active forms of nicotinic acid
  • Found in living cells primarily in the oxidized state
  • Serves as a coenzyme of the dehydrogenases, especially in the dehydrogenation of primary and secondary alcohols
  • NAD usually acts as a hydrogen acceptor, forming NADH which then serves as a hydrogen donor in the respiratory chain
  • Coenzyme for hydride-transfer enzymes and a substrate for NAD(+)-consuming enzymes
  • Recently reported to be a P2Y11 agonist
  • β-Nicotinamide adenine dinucleotide is a cofactor for alcohol dehydrogenase that serves as inhibitory neurotransmitter in visceral smooth muscle
TRUSTED_SUSTAINABILITY

Specifications

Color White
pH 2 to 4 (1% aq. soln.)
Packaging Amber Glass Bottle
Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.