Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

β-Nicotinamide adenine dinucleotide trihydrate, 98%, Thermo Scientific™

Catalog No. AC415620050
Change view
Click to view available options
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC415620050 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC415620050 Supplier Thermo Scientific Chemicals Supplier No. 415620050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1082604-19-0, 53-84-9
Absorbance lambda max.: 259 +/- 2nm (pH 7.0), molar absorptivity: 18000 (260nm, pH 7.0), 0.8 to 0.86 (A250/A260), 0.19 to 0.25 (A280/A260)
CAS Min % 97.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C21H27N7O14P2·3H2O
Quantity 5g
Formula Weight 717.47
Percent Purity ≥97.5% (enzymatic) (on dry substance)
Grade Biochemical
Chemical Name or Material β-Nicotinamide adenine dinucleotide trihydrate, For biochemistry
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.