Learn More
Cayman Chemical Nicotinamide-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 347841-88-7, 10 mg
SDP
Catalog No. 502947242
Encompass_Preferred
An internal standard for the quantification of nicotinamide by GC- or LC-MS
Catalog No. 50-294-7242 Supplier Cayman Chemical Supplier No. 3444610 MG
An internal standard for the quantification of nicotinamide by GC- or LC-MS