Learn More
Medchemexpress LLC Nicotinamide-d4 | 347841-88-7 | 100.0% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB0150S
Encompass_Preferred
Nicotinamide-d4 is the deuterium labeled version of Nicotinamide, a form of vitamin B3 essential for cell physiology. It facilitates NAD+ redox homeostasis, provides NAD+ as a substrate for enzymes in non-redox reactions, and acts as an inhibitor of SIRT1. This product is intended for research use only.
  • Used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration can potentially influence the pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-06520