Learn More
Medchemexpress LLC Nicotinamide-d4 | 347841-88-7 | 99.9% | 1 MG
SDP
Catalog No. 5000406305
Encompass_Preferred
Nicotinamide-d4 is the deuterium labeled Nicotinamide, a form of vitamin B3 essential for cell physiology through NAD+ redox homeostasis. It serves as a substrate for enzymes catalyzing non-redox reactions and is an inhibitor of SIRT1. This product is intended for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-06305 Supplier Medchemexpress LLC Supplier No. HYB0150S1MG
Nicotinamide-d4 is the deuterium labeled Nicotinamide, a form of vitamin B3 essential for cell physiology through NAD+ redox homeostasis. It serves as a substrate for enzymes catalyzing non-redox reactions and is an inhibitor of SIRT1. This product is intended for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS