Learn More
Cayman Chemical Nicotinamide-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 347841-88-7, 5 mg
SDP
Catalog No. 502949564
Encompass_Preferred
An internal standard for the quantification of nicotinamide by GC- or LC-MS
Catalog No. 50-294-9564 Supplier Cayman Chemical Supplier No. 344465 MG
An internal standard for the quantification of nicotinamide by GC- or LC-MS