Learn More
Medchemexpress LLC Nicotinamide-guanine dinucleotide sodium | 2600354-60-5 | 701.41 | 5 MG
SDP
Catalog No. 5000392044
Encompass_Preferred
Nicotinamide-guanine dinucleotide sodium, a NAD sodium (HY-B0445A) analog, is an oxidized form of nicotinamide guanine dinucleotide. It serves as a coenzyme for alcohol dehydrogenase (ADH) in vitro.
  • Serves as coenzymes for alcohol dehydrogenase (ADH) in vitro
Catalog No. 50-003-92044 Supplier Medchemexpress LLC Supplier No. HY164690A5MG
Nicotinamide-guanine dinucleotide sodium, a NAD sodium (HY-B0445A) analog, is an oxidized form of nicotinamide guanine dinucleotide. It serves as a coenzyme for alcohol dehydrogenase (ADH) in vitro.
  • Serves as coenzymes for alcohol dehydrogenase (ADH) in vitro