Learn More
Medchemexpress LLC Nicotinamide-guanine dinucleotide sodium | 2600354-60-5 | 701.41 | C21H26N7NaO15P2 | 10 MG
SDP
Catalog No. 5000289280
Encompass_Preferred
Nicotinamide-guanine dinucleotide sodium is a sodium salt analog of nicotinamide-guanine dinucleotide (an NAD analog) supplied as a research reagent. It functions as a coenzyme for alcohol dehydrogenase (ADH) in vitro and is provided in small laboratory pack sizes.
  • Functions as a coenzyme for alcohol dehydrogenase in vitro.
  • Sodium salt form suitable for biochemical assays.
  • Molecular weight 701.41 and formula C21H26N7NaO15P2.
  • Provided in a 10 mg research pack for laboratory use.
  • Intended for research applications and enzymatic studies.
Catalog No. 50-002-89280 Supplier Medchemexpress LLC Supplier No. HY164690A10MG
Nicotinamide-guanine dinucleotide sodium is a sodium salt analog of nicotinamide-guanine dinucleotide (an NAD analog) supplied as a research reagent. It functions as a coenzyme for alcohol dehydrogenase (ADH) in vitro and is provided in small laboratory pack sizes.
  • Functions as a coenzyme for alcohol dehydrogenase in vitro.
  • Sodium salt form suitable for biochemical assays.
  • Molecular weight 701.41 and formula C21H26N7NaO15P2.
  • Provided in a 10 mg research pack for laboratory use.
  • Intended for research applications and enzymatic studies.