Learn More
Medchemexpress LLC β-Nicotinamide mononucleotide, reduced form disodium | 108347-85-9 | 99.6% | 380.20 | C11H15N2Na2O8P | 10 MG
SDP
Catalog No. 5000284364
Encompass_Preferred
β-Nicotinamide mononucleotide, reduced form disodium (NMNH) is the reduced form of β-nicotinamide mononucleotide and an orally active NAD+ precursor. It increases cellular NAD+ and NADH levels and is used in research to study metabolic pathways and cell growth. Supplied as a light yellow solid with high purity and recommended storage under inert atmosphere to preserve stability.
  • Orally active NAD+ precursor that raises cellular NAD+/NADH levels.
  • Reduced form of nicotinamide mononucleotide with distinct metabolic effects.
  • Reported to inhibit glycolysis and affect the tricarboxylic acid cycle and cell growth.
  • High purity suitable for research applications.
  • Light yellow solid; store protected from light under nitrogen at -20°C.
Catalog No. 50-002-84364 Supplier Medchemexpress LLC Supplier No. HYW615108B10MG
β-Nicotinamide mononucleotide, reduced form disodium (NMNH) is the reduced form of β-nicotinamide mononucleotide and an orally active NAD+ precursor. It increases cellular NAD+ and NADH levels and is used in research to study metabolic pathways and cell growth. Supplied as a light yellow solid with high purity and recommended storage under inert atmosphere to preserve stability.
  • Orally active NAD+ precursor that raises cellular NAD+/NADH levels.
  • Reduced form of nicotinamide mononucleotide with distinct metabolic effects.
  • Reported to inhibit glycolysis and affect the tricarboxylic acid cycle and cell growth.
  • High purity suitable for research applications.
  • Light yellow solid; store protected from light under nitrogen at -20°C.