Learn More
Medchemexpress LLC β-nicotinamide mononucleotide, reduced form disodium | 108347-85-9 | 99.6% | 380.20 | C11H15N2Na2O8P | 50 MG
SDP
Catalog No. 5000283919
Encompass_Preferred
β-nicotinamide mononucleotide, reduced form disodium (NMNH) is the reduced form of nicotinamide mononucleotide supplied as a disodium salt for research use. It is reported to elevate intracellular NAD+ levels and modulate cellular metabolism in vitro and in vivo. The product is supplied as a light yellow to yellow solid with supporting analytical data (HPLC, NMR, MS) and a certificate of analysis for purity and identity.
  • Reduced form of nicotinamide mononucleotide with disodium counterions.
  • Reported to increase intracellular NAD+ levels in cellular and animal studies.
  • Analytically verified by HPLC, 1H NMR, and mass spectrometry.
  • Available in multiple milligram pack sizes to support dosing studies.
  • Supplied as a solid with storage recommendations to preserve stability.
Catalog No. 50-002-83919 Supplier Medchemexpress LLC Supplier No. HYW615108B50MG
β-nicotinamide mononucleotide, reduced form disodium (NMNH) is the reduced form of nicotinamide mononucleotide supplied as a disodium salt for research use. It is reported to elevate intracellular NAD+ levels and modulate cellular metabolism in vitro and in vivo. The product is supplied as a light yellow to yellow solid with supporting analytical data (HPLC, NMR, MS) and a certificate of analysis for purity and identity.
  • Reduced form of nicotinamide mononucleotide with disodium counterions.
  • Reported to increase intracellular NAD+ levels in cellular and animal studies.
  • Analytically verified by HPLC, 1H NMR, and mass spectrometry.
  • Available in multiple milligram pack sizes to support dosing studies.
  • Supplied as a solid with storage recommendations to preserve stability.