Learn More
Medchemexpress LLC β-nicotinamide mononucleotide, reduced form disodium | 108347-85-9 | 99.6% | 380.20 g·mol-1 | C11H15N2Na2O8P | 25 MG
SDP
Catalog No. 5000284068
Encompass_Preferred
β-Nicotinamide mononucleotide, reduced form (disodium) is the reduced form of β-nicotinamide mononucleotide (NMNH), an endogenous NAD+ precursor used in biochemical and cellular research. The disodium salt is supplied as a solid and is characterized for identity and purity.
  • Provides the reduced NMN (NMNH) disodium salt for NAD+ metabolism studies.
  • High purity suitable for analytical and cellular applications.
  • Solid form with light yellow to yellow appearance.
  • Storage: -20°C for solid; in solvent: -80°C up to 6 months or -20°C up to 1 month.
  • Characterized by HPLC, 1H NMR, and MS to confirm identity.
Catalog No. 50-002-84068 Supplier Medchemexpress LLC Supplier No. HYW615108B25MG
β-Nicotinamide mononucleotide, reduced form (disodium) is the reduced form of β-nicotinamide mononucleotide (NMNH), an endogenous NAD+ precursor used in biochemical and cellular research. The disodium salt is supplied as a solid and is characterized for identity and purity.
  • Provides the reduced NMN (NMNH) disodium salt for NAD+ metabolism studies.
  • High purity suitable for analytical and cellular applications.
  • Solid form with light yellow to yellow appearance.
  • Storage: -20°C for solid; in solvent: -80°C up to 6 months or -20°C up to 1 month.
  • Characterized by HPLC, 1H NMR, and MS to confirm identity.