Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC β-nicotinamide mononucleotide, reduced form disodium | 108347-85-9 | 99.6% | 380.20 g/mol | C11H15N2Na2O8P | 5 MG
SDP

Catalog No. 5000284066
Encompass_Preferred

β-Nicotinamide mononucleotide, reduced form disodium (NMNH) is the reduced disodium salt of nicotinamide mononucleotide supplied for research use in studies of NAD+ metabolism. It is provided as a powder with available analytical data for identity and purity and recommended cold, light-protected storage.

  • Chemical formula: C11H15N2Na2O8P.
  • Molecular weight: 380.20 g/mol.
  • CAS number: 108347-85-9.
  • Purity: 99.6% by HPLC.
  • Recommended storage: protect from light, store under nitrogen at -20°C; in solution store at -80°C for long term.
  • Analytical data available: COA, H NMR, C NMR, RP-HPLC, MS.
  • Typical pack sizes: 1 mg, 5 mg, 10 mg.

Catalog No. 50-002-84066 Supplier Medchemexpress LLC Supplier No. HYW615108B5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

β-Nicotinamide mononucleotide, reduced form disodium (NMNH) is the reduced disodium salt of nicotinamide mononucleotide supplied for research use in studies of NAD+ metabolism. It is provided as a powder with available analytical data for identity and purity and recommended cold, light-protected storage.

  • Chemical formula: C11H15N2Na2O8P.
  • Molecular weight: 380.20 g/mol.
  • CAS number: 108347-85-9.
  • Purity: 99.6% by HPLC.
  • Recommended storage: protect from light, store under nitrogen at -20°C; in solution store at -80°C for long term.
  • Analytical data available: COA, H NMR, C NMR, RP-HPLC, MS.
  • Typical pack sizes: 1 mg, 5 mg, 10 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.