Learn More
Medchemexpress LLC Nicotinamide N-Methy 100ug | 100ug
SDP
Catalog No. 5000194747
Encompass_Preferred
The nicotinamide N-methyltransferase (NNMT) protein catalyzes the methylation of nicotinamide using S-adenosyl-L-methionine to form N1-methylnicotinamide NNMT affects pluripotent embryonic stem cell development and acts as a metabolic regulator affecting adipose tissue energy expenditure gluconeogenesis and cholesterol biosynthesis Nicotinamide N-Methyltransferase/NNMT Protein Human (His) is the recombinant human-derived Nicotinamide N-Methyltransferase/NNMT protein expressed by E coli with C-His labeled tag
Catalog No. 50-001-94747 Supplier Medchemexpress LLC Supplier No. HYP79286100UG
The nicotinamide N-methyltransferase (NNMT) protein catalyzes the methylation of nicotinamide using S-adenosyl-L-methionine to form N1-methylnicotinamide NNMT affects pluripotent embryonic stem cell development and acts as a metabolic regulator affecting adipose tissue energy expenditure gluconeogenesis and cholesterol biosynthesis Nicotinamide N-Methyltransferase/NNMT Protein Human (His) is the recombinant human-derived Nicotinamide N-Methyltransferase/NNMT protein expressed by E coli with C-His labeled tag