Learn More
Medchemexpress LLC HY-101407 100mg Medchemexpress, Nicotinamide N-oxide CAS:1986-81-8 Purity:>98%
SDP
Catalog No. 502080632
Encompass_Preferred
Medchemexpress, HY-101407 100mg Nicotinamide N-oxide CAS:1986-81-8 Nicotinamide N-oxide, an in vivo nicotinamide metabolite, is a potent, and selective antagonist of the CXCR2 receptor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0632 Supplier Medchemexpress LLC Supplier No. HY101407100MG
Add to Cart
Add to Cart
Medchemexpress, HY-101407 100mg Nicotinamide N-oxide CAS:1986-81-8 Nicotinamide N-oxide, an in vivo nicotinamide metabolite, is a potent, and selective antagonist of the CXCR2 receptor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.