Learn More
Cayman Chemical Nicotinamide N-oxide, Purity: ≥95%, CAS: 1986-81-8, 50 g
SDP
Catalog No. 502939781
Encompass_Preferred
A metabolite of nicotinamide; formed via oxidation of nicotinamide by CYP2E1; inhibits proliferation and induces reduction of NBT in HL-60 promyelocytic leukemia cells at 30 mM; levels in urine are elevated in a mouse model of high-fat diet-induced obesity
Catalog No. 50-293-9781 Supplier Cayman Chemical Supplier No. 2844150 G
A metabolite of nicotinamide; formed via oxidation of nicotinamide by CYP2E1; inhibits proliferation and induces reduction of NBT in HL-60 promyelocytic leukemia cells at 30 mM; levels in urine are elevated in a mouse model of high-fat diet-induced obesity