Learn More
Medchemexpress LLC Nicotinamide (Niacinamide) | 98-92-0 | 99.99% | 100 MG
SDP
Catalog No. 5000406304
Encompass_Preferred
Nicotinamide is a form of vitamin B3 (niacin) that inhibits SIRT2 and SIRT1 activity. It increases cellular NAD+, ATP, and ROS levels, and is known to inhibit tumor growth, improve survival, and exhibit anti-HBV activity. It is intended for research use only.
  • Inhibits SIRT2 and SIRT1 activity
  • Increases cellular NAD+, ATP, and ROS levels
  • Inhibits tumor growth and improves survival
  • Possesses anti-HBV activity
  • Reduces cell number in A375 and SK-MEL-28 cells
  • Promotes pancreatic cell differentiation from human embryonic stem cells
Catalog No. 50-004-06304 Supplier Medchemexpress LLC Supplier No. HYB0150R100MG
Nicotinamide is a form of vitamin B3 (niacin) that inhibits SIRT2 and SIRT1 activity. It increases cellular NAD+, ATP, and ROS levels, and is known to inhibit tumor growth, improve survival, and exhibit anti-HBV activity. It is intended for research use only.
  • Inhibits SIRT2 and SIRT1 activity
  • Increases cellular NAD+, ATP, and ROS levels
  • Inhibits tumor growth and improves survival
  • Possesses anti-HBV activity
  • Reduces cell number in A375 and SK-MEL-28 cells
  • Promotes pancreatic cell differentiation from human embryonic stem cells