Learn More
Medkoo Biosciences Inc Nicotinamide Riboside Chloride, 23111-00-4, MFCD27952781, 1g
SDP
Catalog No. NC1611436
Encompass
Chemical Formula: C11H15ClN2O5, Purity:>98%, Molecular Weight: 290.7, Soluble in DMSO and water (10mg/mL). Nicotinamide riboside, also known as NR and SRT647, is a pyridine-nucleoside form of vitamin B3 that functions as a precursor to nicotinamide adenine dinucleotide or NAD+. NR blocks degeneration of surgically severed dorsal root ganglion neurons ex vivo and protects against noise-induced hearing loss in living mice. Nicotinamide riboside prevents muscle, neural and melanocyte stem cell senescence
Catalog No. NC1611436 Supplier Medkoo Biosciences Inc Supplier No. 3294791G
Chemical Formula: C11H15ClN2O5, Purity:>98%, Molecular Weight: 290.7, Soluble in DMSO and water (10mg/mL). Nicotinamide riboside, also known as NR and SRT647, is a pyridine-nucleoside form of vitamin B3 that functions as a precursor to nicotinamide adenine dinucleotide or NAD+. NR blocks degeneration of surgically severed dorsal root ganglion neurons ex vivo and protects against noise-induced hearing loss in living mice. Nicotinamide riboside prevents muscle, neural and melanocyte stem cell senescence