Learn More
TARGETMOL CHEMICALS INC NICOTINAMIDE RIBOSIDE 200MG
SDP
Catalog No. 502581327
Encompass_Preferred
Also available in 5 mg and bulk. Please contact Fisher for quotes. Nicotinamide riboside increases NAD[+] levels and activates SIRT1 and SIRT3 culminating in enhanced oxidative metabolism and protection against high fat diet-induced metabolic abnormalities. purity: 99%
Catalog No. 50-258-1327 Supplier TARGETMOL CHEMICALS INC Supplier No. T13795200MG
Also available in 5 mg and bulk. Please contact Fisher for quotes. Nicotinamide riboside increases NAD[+] levels and activates SIRT1 and SIRT3 culminating in enhanced oxidative metabolism and protection against high fat diet-induced metabolic abnormalities. purity: 99%