Learn More
Cayman Chemical Nicotinamide riboside, Purity: ≥95% Contains ~80% cyclodextrin as stabilizer, CAS: 1341-23-7, 25 mg
SDP
Catalog No. 502994006
Encompass_Preferred
A riboside form of nicotinamide; a precursor of NAD+ and a source of vitamin B3; increases intracellular and mitochondrial NAD+ content in C2C12, Hepa1.6, and HEK293 cells in a concentration-dependent manner at concentrations ranging from 1-1,000 μM; decreases acetylation of FOXO1 and SOD2 but not tubulin; increases NAD+ levels in liver and skeletal muscle and prevents body weight gain in mice fed a high-fat diet when used at a dose of 400 mg/kg per day; increases NAD+ in the cerebral cortex and reduces cognitive deterioration in a transgenic mouse model of Alzheimer’s disease
Catalog No. 50-299-4006 Supplier Cayman Chemical Supplier No. 2313225 MG
A riboside form of nicotinamide; a precursor of NAD+ and a source of vitamin B3; increases intracellular and mitochondrial NAD+ content in C2C12, Hepa1.6, and HEK293 cells in a concentration-dependent manner at concentrations ranging from 1-1,000 μM; decreases acetylation of FOXO1 and SOD2 but not tubulin; increases NAD+ levels in liver and skeletal muscle and prevents body weight gain in mice fed a high-fat diet when used at a dose of 400 mg/kg per day; increases NAD+ in the cerebral cortex and reduces cognitive deterioration in a transgenic mouse model of Alzheimer’s disease