Learn More
Medchemexpress LLC HY-123033A 100mg Medchemexpress, Nicotinamide riboside (chloride) CAS:23111-00-4 Purity:>98%
SDP
Catalog No. 501932575
Encompass_Preferred
Medchemexpress, HY-123033A 100mg Nicotinamide riboside (chloride) CAS:23111-00-4 Nicotinamide riboside chloride is a crystal form of Nicotinamide riboside (NR) chloride. Nicotinamide riboside chloride increases NAD [+] levels and activates SIRT1 and SIRT3 , culminating in enhanced oxidative metabolism and protection against high fat diet-induced metabolic abnormalities. Nicotinamide riboside chloride is used in dietary supplements [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2575 Supplier Medchemexpress LLC Supplier No. HY123033A100MG
Medchemexpress, HY-123033A 100mg Nicotinamide riboside (chloride) CAS:23111-00-4 Nicotinamide riboside chloride is a crystal form of Nicotinamide riboside (NR) chloride. Nicotinamide riboside chloride increases NAD [+] levels and activates SIRT1 and SIRT3 , culminating in enhanced oxidative metabolism and protection against high fat diet-induced metabolic abnormalities. Nicotinamide riboside chloride is used in dietary supplements [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.