Learn More
Sigma Aldrich Fine Chemicals Biosciences Nicotinamide riboside chlo50MG
Catalog No. 502899592
Encompass
Nicotinamide riboside is involved in nicotinate and nicotinamide metabolism. Nicotinamide riboside was originally identified as a nutrient in milk and later discovered to be a NAD() precursor.
Catalog No. 50-289-9592 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SMB0090750MG
Nicotinamide riboside is involved in nicotinate and nicotinamide metabolism. Nicotinamide riboside was originally identified as a nutrient in milk and later discovered to be a NAD() precursor.