Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nicotinamide riboside chloride | 23111-00-4 | MFCD27952781 | 99.9% | 290.70 | C11H15ClN2O5 | 10G
SDP

Catalog No. 5000238709
Encompass_Preferred

Nicotinamide riboside chloride is the chloride salt of nicotinamide riboside, a bioactive NAD+ precursor commonly used in metabolic and sirtuin-related research. Supplied as a white to light yellow solid powder with high purity, it is suitable for in vitro and in vivo studies and dissolves in common laboratory solvents with ultrasonic assistance.

  • High purity (99.9%) solid powder suitable for research use.
  • Soluble in water, DMSO, and PBS; ultrasonic assistance recommended.
  • Provides a precursor to NAD+ for metabolic and sirtuin studies.
  • Available in multiple pack sizes including 10 G for larger experiments.
  • Molecular weight 290.70 and formula C11H15ClN2O5 for reference.

Catalog No. 50-002-38709 Supplier Medchemexpress LLC Supplier No. HY123033A10G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nicotinamide riboside chloride is the chloride salt of nicotinamide riboside, a bioactive NAD+ precursor commonly used in metabolic and sirtuin-related research. Supplied as a white to light yellow solid powder with high purity, it is suitable for in vitro and in vivo studies and dissolves in common laboratory solvents with ultrasonic assistance.

  • High purity (99.9%) solid powder suitable for research use.
  • Soluble in water, DMSO, and PBS; ultrasonic assistance recommended.
  • Provides a precursor to NAD+ for metabolic and sirtuin studies.
  • Available in multiple pack sizes including 10 G for larger experiments.
  • Molecular weight 290.70 and formula C11H15ClN2O5 for reference.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.