Learn More
Medchemexpress LLC Nicotinamide riboside chloride | 23111-00-4 | MFCD27952781 | 99.9% | 290.70 | C11H15ClN2O5 | 1G
SDP
Catalog No. 5000236324
Encompass_Preferred
Nicotinamide riboside chloride is a nicotinamide (vitamin B3) riboside salt and an NAD+ precursor commonly used in metabolic and aging research to elevate intracellular NAD+ and modulate sirtuin activity. It is supplied as a high-purity solid and as a DMSO solution for use in biochemical and cell-based assays, and is provided with supporting documentation such as a COA and SDS.
  • Increases intracellular NAD+ levels.
  • Activates sirtuin enzymes such as SIRT1 and SIRT3.
  • Available as solid and DMSO solution for assay flexibility.
  • High purity suitable for biochemical and cell-based studies.
  • Provided with certificate of analysis and safety data sheet.
Catalog No. 50-002-36324 Supplier Medchemexpress LLC Supplier No. HY123033A1G
Nicotinamide riboside chloride is a nicotinamide (vitamin B3) riboside salt and an NAD+ precursor commonly used in metabolic and aging research to elevate intracellular NAD+ and modulate sirtuin activity. It is supplied as a high-purity solid and as a DMSO solution for use in biochemical and cell-based assays, and is provided with supporting documentation such as a COA and SDS.
  • Increases intracellular NAD+ levels.
  • Activates sirtuin enzymes such as SIRT1 and SIRT3.
  • Available as solid and DMSO solution for assay flexibility.
  • High purity suitable for biochemical and cell-based studies.
  • Provided with certificate of analysis and safety data sheet.