Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nicotinamide riboside chloride | 23111-00-4 | MFCD27952781 | 100.0% | 290.70 | C11H15ClN2O5 | 5MG
SDP

Catalog No. 5000238535
Encompass_Preferred

Nicotinamide riboside chloride (standard) is an analytical standard of nicotinamide riboside chloride used for research and analytical applications. It is a high-purity NAD+ precursor and source of vitamin B3, supplied as a solid for quantitation, method validation, and metabolic studies.

  • High purity suitable for analytical and research use.
  • Available in multiple small sizes for use as standards.
  • CAS 23111-00-4 provides unambiguous compound identification.
  • Molecular weight and formula provided for accurate calculations.
  • Applicable in studies of NAD+ metabolism and sirtuin activation.

Catalog No. 50-002-38535 Supplier Medchemexpress LLC Supplier No. HY123033AR5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nicotinamide riboside chloride (standard) is an analytical standard of nicotinamide riboside chloride used for research and analytical applications. It is a high-purity NAD+ precursor and source of vitamin B3, supplied as a solid for quantitation, method validation, and metabolic studies.

  • High purity suitable for analytical and research use.
  • Available in multiple small sizes for use as standards.
  • CAS 23111-00-4 provides unambiguous compound identification.
  • Molecular weight and formula provided for accurate calculations.
  • Applicable in studies of NAD+ metabolism and sirtuin activation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.