Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nicotinamide riboside chloride | 23111-00-4 | MFCD27952781 | 99.9% | 290.70 g·mol⁻1 | C11H15ClN2O5 | 50G
SDP

Catalog No. 5000237908
Encompass_Preferred

Nicotinamide riboside chloride is an NAD+ precursor used in research to elevate cellular NAD+ levels and modulate sirtuin activity. Supplied as a powder, it is intended for biochemical and metabolic research applications including aging and neurodegeneration studies.

  • High purity: 99.9%.
  • Form: powder suitable for analytical and biochemical use.
  • Molecular formula: C11H15ClN2O5; molecular weight: 290.70 g·mol⁻1.
  • Application: used in NAD+ biosynthesis and sirtuin activation studies.
  • Packaging: available as a 50 g research pack.

Catalog No. 50-002-37908 Supplier Medchemexpress LLC Supplier No. HY123033A50G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nicotinamide riboside chloride is an NAD+ precursor used in research to elevate cellular NAD+ levels and modulate sirtuin activity. Supplied as a powder, it is intended for biochemical and metabolic research applications including aging and neurodegeneration studies.

  • High purity: 99.9%.
  • Form: powder suitable for analytical and biochemical use.
  • Molecular formula: C11H15ClN2O5; molecular weight: 290.70 g·mol⁻1.
  • Application: used in NAD+ biosynthesis and sirtuin activation studies.
  • Packaging: available as a 50 g research pack.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.