Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nicotinamide riboside chloride | 23111-00-4 | MFCD27952781 | 100.0% | 290.70 g/mol | C11H15ClN2O5 | 10MG
SDP

Catalog No. 5000237336
Encompass_Preferred

Nicotinamide riboside chloride is the chloride salt of nicotinamide riboside, a bioavailable precursor of nicotinamide adenine dinucleotide (NAD+) used in biochemical and cell biology research to elevate cellular NAD+ levels and probe NAD+-dependent pathways. It is supplied as a high-purity, crystalline white to off-white solid suitable for in vitro and analytical applications.

  • High purity: 99.97%.
  • CAS number: 23111-00-4.
  • Molecular formula: C11H15ClN2O5; molecular weight: 290.70 g/mol.
  • Chloride salt form for improved stability and handling.
  • Suitable for studies that increase cellular NAD+ and investigate metabolic or sirtuin-related pathways.
  • Stable solid form for storage and analytical use.

Catalog No. 50-002-37336 Supplier Medchemexpress LLC Supplier No. HY123033AR10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nicotinamide riboside chloride is the chloride salt of nicotinamide riboside, a bioavailable precursor of nicotinamide adenine dinucleotide (NAD+) used in biochemical and cell biology research to elevate cellular NAD+ levels and probe NAD+-dependent pathways. It is supplied as a high-purity, crystalline white to off-white solid suitable for in vitro and analytical applications.

  • High purity: 99.97%.
  • CAS number: 23111-00-4.
  • Molecular formula: C11H15ClN2O5; molecular weight: 290.70 g/mol.
  • Chloride salt form for improved stability and handling.
  • Suitable for studies that increase cellular NAD+ and investigate metabolic or sirtuin-related pathways.
  • Stable solid form for storage and analytical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.