Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nicotinamide riboside chloride | 23111-00-4 | MFCD27952781 | 99.9% | 290.70 g/mol | C11H15ClN2O5 | 10MM 1ML
SDP

Catalog No. 5000237337
Encompass_Preferred

Nicotinamide riboside chloride is a nicotinamide adenine dinucleotide (NAD+) precursor used in biochemical and cellular research to elevate intracellular NAD+ and to probe metabolic and sirtuin-related pathways. It is provided as a ready-to-use 10 mM solution in DMSO and as multiple solid pack sizes to suit assay development and storage needs.

  • High purity (99.9%).
  • Ready-to-use 10 mM solution in DMSO, 1 mL.
  • Available in multiple solid sizes for bulk use or small-scale assays.
  • Molecular formula: C11H15ClN2O5; molecular weight: 290.70 g/mol.
  • Useful as an NAD+ precursor in metabolic and sirtuin activation studies.
  • Stable when stored sealed and protected from moisture and light.

Catalog No. 50-002-37337 Supplier Medchemexpress LLC Supplier No. HY123033A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nicotinamide riboside chloride is a nicotinamide adenine dinucleotide (NAD+) precursor used in biochemical and cellular research to elevate intracellular NAD+ and to probe metabolic and sirtuin-related pathways. It is provided as a ready-to-use 10 mM solution in DMSO and as multiple solid pack sizes to suit assay development and storage needs.

  • High purity (99.9%).
  • Ready-to-use 10 mM solution in DMSO, 1 mL.
  • Available in multiple solid sizes for bulk use or small-scale assays.
  • Molecular formula: C11H15ClN2O5; molecular weight: 290.70 g/mol.
  • Useful as an NAD+ precursor in metabolic and sirtuin activation studies.
  • Stable when stored sealed and protected from moisture and light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.