Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nicotinamide riboside chloride | 23111-00-4 | MFCD27952781 | 99.9% | 290.70 g/mol | C11H15ClN2O5 | 25G
SDP

Catalog No. 5000237907
Encompass_Preferred

Nicotinamide riboside chloride is the chloride salt form of nicotinamide riboside, a pyridine nucleoside and precursor to nicotinamide adenine dinucleotide (NAD+). It is supplied for research use to raise cellular NAD+ levels and support studies of metabolism, mitochondrial function, and aging-related pathways.

  • Precursor to NAD+ for cellular metabolism studies.
  • High purity suitable for research applications.
  • Provided as a solid for accurate dosing and formulation.
  • Molecular weight 290.70 g/mol and chemical formula C11H15ClN2O5.
  • Available in gram-scale pack sizes for laboratory use.

Catalog No. 50-002-37907 Supplier Medchemexpress LLC Supplier No. HY123033A25G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nicotinamide riboside chloride is the chloride salt form of nicotinamide riboside, a pyridine nucleoside and precursor to nicotinamide adenine dinucleotide (NAD+). It is supplied for research use to raise cellular NAD+ levels and support studies of metabolism, mitochondrial function, and aging-related pathways.

  • Precursor to NAD+ for cellular metabolism studies.
  • High purity suitable for research applications.
  • Provided as a solid for accurate dosing and formulation.
  • Molecular weight 290.70 g/mol and chemical formula C11H15ClN2O5.
  • Available in gram-scale pack sizes for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.