Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nicotinamide riboside chloride | 23111-00-4 | MFCD27952781 | 99.9% | 290.70 g/mol | C11H15ClN2O5 | 25MG
SDP

Catalog No. 5000235524
Encompass_Preferred

Nicotinamide riboside chloride is an NAD+ precursor used as a research reagent to increase intracellular NAD+ levels and activate sirtuin enzymes (SIRT1, SIRT3). It is used in metabolic, mitochondrial, and neurodegenerative disease studies to probe NAD+ metabolism, oxidative metabolism, and neuroprotection.

  • High purity suitable for research applications (99.9%).
  • Orally active NAD+ precursor that elevates intracellular NAD+ and modulates sirtuin activity.
  • Commonly used to study oxidative metabolism, mitochondrial function, and neuroprotection.
  • Chemical formula C11H15ClN2O5; molecular weight 290.70 g/mol.
  • White to light-yellow solid, readily soluble in DMSO.
  • Available in small research packs (e.g., 25 mg).

Catalog No. 50-002-35524 Supplier Medchemexpress LLC Supplier No. HY123033AR25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nicotinamide riboside chloride is an NAD+ precursor used as a research reagent to increase intracellular NAD+ levels and activate sirtuin enzymes (SIRT1, SIRT3). It is used in metabolic, mitochondrial, and neurodegenerative disease studies to probe NAD+ metabolism, oxidative metabolism, and neuroprotection.

  • High purity suitable for research applications (99.9%).
  • Orally active NAD+ precursor that elevates intracellular NAD+ and modulates sirtuin activity.
  • Commonly used to study oxidative metabolism, mitochondrial function, and neuroprotection.
  • Chemical formula C11H15ClN2O5; molecular weight 290.70 g/mol.
  • White to light-yellow solid, readily soluble in DMSO.
  • Available in small research packs (e.g., 25 mg).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.