Learn More
Medchemexpress LLC Nicotinamide riboside chloride | 23111-00-4 | MFCD27952781 | 99.9% | 290.70 g/mol | C11H15ClN2O5 | 500MG
SDP
Catalog No. 5000237335
Encompass_Preferred
Nicotinamide riboside chloride is the chloride salt of nicotinamide riboside, an NAD+ precursor used in metabolic and neuroprotection research. It elevates intracellular NAD+ levels and activates sirtuin enzymes, supporting studies of oxidative metabolism, energy homeostasis, and preclinical models of metabolic or cognitive dysfunction. Supplied as a high-purity research chemical for biochemical and cell-based assays.
  • High purity suitable for biochemical and cell-based assays.
  • Chloride salt form for improved stability and solubility.
  • Acts as an NAD+ precursor and sirtuin activator.
  • Applicable to metabolic, oxidative stress, and neuroprotection studies.
  • Available in multiple pack sizes, including 500 mg.
Catalog No. 50-002-37335 Supplier Medchemexpress LLC Supplier No. HY123033A500MG
Nicotinamide riboside chloride is the chloride salt of nicotinamide riboside, an NAD+ precursor used in metabolic and neuroprotection research. It elevates intracellular NAD+ levels and activates sirtuin enzymes, supporting studies of oxidative metabolism, energy homeostasis, and preclinical models of metabolic or cognitive dysfunction. Supplied as a high-purity research chemical for biochemical and cell-based assays.
  • High purity suitable for biochemical and cell-based assays.
  • Chloride salt form for improved stability and solubility.
  • Acts as an NAD+ precursor and sirtuin activator.
  • Applicable to metabolic, oxidative stress, and neuroprotection studies.
  • Available in multiple pack sizes, including 500 mg.