Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nicotinamide riboside chloride | 23111-00-4 | MFCD27952781 | 100.0% | 290.70 g/mol | C11H15ClN2O5 | 50MG
SDP

Catalog No. 5000237577
Encompass_Preferred

Nicotinamide riboside chloride is a high-purity analytical reference standard supplied as a white to off-white solid for analytical and laboratory research applications. It is provided in small pack sizes to support accurate handling and reference use.

  • Purity 99.97%.
  • Molecular weight 290.70 g/mol.
  • Molecular formula C11H15ClN2O5.
  • Supplied as a solid, white to off-white.
  • COA and SDS available for quality and safety data.
  • Pack size 50 MG suitable for analytical use.
  • Shipping typically at room temperature; follow COA/SDS for storage.

Catalog No. 50-002-37577 Supplier Medchemexpress LLC Supplier No. HY123033AR50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nicotinamide riboside chloride is a high-purity analytical reference standard supplied as a white to off-white solid for analytical and laboratory research applications. It is provided in small pack sizes to support accurate handling and reference use.

  • Purity 99.97%.
  • Molecular weight 290.70 g/mol.
  • Molecular formula C11H15ClN2O5.
  • Supplied as a solid, white to off-white.
  • COA and SDS available for quality and safety data.
  • Pack size 50 MG suitable for analytical use.
  • Shipping typically at room temperature; follow COA/SDS for storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.