Learn More
Medchemexpress LLC Nicotinamide riboside malate | 2415659-01-5 | MFCD34549739 | 98.0% | 388.33 g/mol | C15H20N2O10 | 1G
SDP
Catalog No. 5000237912
Encompass_Preferred
Nicotinamide riboside malate is the malate salt form of nicotinamide riboside, a bioavailable precursor of nicotinamide adenine dinucleotide (NAD+) used in research to elevate intracellular NAD+ levels and probe sirtuin-mediated pathways. It is commonly employed in studies of metabolism, aging, and mitochondrial function and is provided as a solid reagent for laboratory research (CAS 2415659-01-5).
  • Orally bioavailable NAD+ precursor.
  • Supports increased intracellular NAD+ levels.
  • Enables sirtuin activation and metabolic pathway studies.
  • Suitable for research on aging and mitochondrial function.
  • Supplied as a solid powder for laboratory use.
Catalog No. 50-002-37912 Supplier Medchemexpress LLC Supplier No. HY123033C1G
Nicotinamide riboside malate is the malate salt form of nicotinamide riboside, a bioavailable precursor of nicotinamide adenine dinucleotide (NAD+) used in research to elevate intracellular NAD+ levels and probe sirtuin-mediated pathways. It is commonly employed in studies of metabolism, aging, and mitochondrial function and is provided as a solid reagent for laboratory research (CAS 2415659-01-5).
  • Orally bioavailable NAD+ precursor.
  • Supports increased intracellular NAD+ levels.
  • Enables sirtuin activation and metabolic pathway studies.
  • Suitable for research on aging and mitochondrial function.
  • Supplied as a solid powder for laboratory use.