Learn More
Medchemexpress LLC Nicotinamide riboside malate | 2415659-01-5 | MFCD34549739 | 99.0% | 388.33 g/mol | C15H20N2O10 | 500MG
SDP
Catalog No. 5000238791
Encompass_Preferred
Nicotinamide riboside malate is a salt of nicotinamide riboside and malic acid used as an NAD+ precursor and source of vitamin B3. It is employed in biochemical and cellular research to elevate cellular NAD+ levels, probe sirtuin-mediated pathways, and study metabolic and neuroprotective effects in vitro and in vivo models.
  • Orally active NAD+ precursor
  • Raises cellular NAD+ levels
  • Activates sirtuin pathways
  • Useful for metabolic and neuroprotection studies
  • Stable malate salt suitable for biochemical assays
Catalog No. 50-002-38791 Supplier Medchemexpress LLC Supplier No. HY123033C500MG
Nicotinamide riboside malate is a salt of nicotinamide riboside and malic acid used as an NAD+ precursor and source of vitamin B3. It is employed in biochemical and cellular research to elevate cellular NAD+ levels, probe sirtuin-mediated pathways, and study metabolic and neuroprotective effects in vitro and in vivo models.
  • Orally active NAD+ precursor
  • Raises cellular NAD+ levels
  • Activates sirtuin pathways
  • Useful for metabolic and neuroprotection studies
  • Stable malate salt suitable for biochemical assays