Learn More
Cayman Chemical Nicotinamide riboside-d4 (triflate), Purity: ≥95%, 5 mg
SDP
Catalog No. 502948926
Encompass_Preferred
An internal standard for the quantification of nicotinamide riboside by GC- or LC-MS
Catalog No. 50-294-8926 Supplier Cayman Chemical Supplier No. 343115 MG
An internal standard for the quantification of nicotinamide riboside by GC- or LC-MS