Learn More
STEMCELL Technologies Nicotinamide, Size: 100 g
SDP
Catalog No. 501974385
Encompass
Nicotinamide (NAM) is a cell culture supplement used in the differentiation of embryonic stem (ES) and induced pluripotent stem (iPS) cells. It is an amide derivative of vitamin B3, a poly (ADP-ribose) polymerase (PARP) inhibitor, and represents the primary precursor of NAD+. Nicotinamide has also been shown to modulate stem cell differentiation in various applications, most notably for the differentiation of pancreatic mouse ES and iPS cells to pancreatic islet-like insulin-secreting cells.
Catalog No. 50-197-4385 Supplier STEMCELL Technologies Supplier No. 07154
Nicotinamide (NAM) is a cell culture supplement used in the differentiation of embryonic stem (ES) and induced pluripotent stem (iPS) cells. It is an amide derivative of vitamin B3, a poly (ADP-ribose) polymerase (PARP) inhibitor, and represents the primary precursor of NAD+. Nicotinamide has also been shown to modulate stem cell differentiation in various applications, most notably for the differentiation of pancreatic mouse ES and iPS cells to pancreatic islet-like insulin-secreting cells.