Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nicotinamide (Standard) | 98-92-0 | 99.9% | 50 MG
SDP

Catalog No. 5000406223
Encompass_Preferred

Nicotinamide (Standard) is the analytical standard of Nicotinamide, a form of vitamin B3 or niacin. This product is intended for research and analytical applications. Nicotinamide Hydrochloride is known to inhibit SIRT2 activity with an IC50 of 2 μM, and it also inhibits SIRT1. It increases cellular NAD+, ATP, and ROS levels, and has been observed to inhibit tumor growth, improve survival, and demonstrate anti-HBV activity.

  • Application: The compound serves as an analytical standard, commonly utilized in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Molecular weight: 122.12
  • Formula: C6H6N2O
  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Storage: Please store the product under the recommended conditions specified in the Certificate of Analysis.

Catalog No. 50-004-06223 Supplier Medchemexpress LLC Supplier No. HYB0150R50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nicotinamide (Standard) is the analytical standard of Nicotinamide, a form of vitamin B3 or niacin. This product is intended for research and analytical applications. Nicotinamide Hydrochloride is known to inhibit SIRT2 activity with an IC50 of 2 μM, and it also inhibits SIRT1. It increases cellular NAD+, ATP, and ROS levels, and has been observed to inhibit tumor growth, improve survival, and demonstrate anti-HBV activity.

  • Application: The compound serves as an analytical standard, commonly utilized in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Molecular weight: 122.12
  • Formula: C6H6N2O
  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Storage: Please store the product under the recommended conditions specified in the Certificate of Analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.