Learn More
Alkali Scientific Nicotinamide [Vitamin PP], 100 G
SDP
Catalog No. 5000660833
Encompass_Preferred
Nicotinamide (Vitamin PP) is a key biochemical reagent used in a variety of applications related to cellular metabolism and enzymatic activity. This 100g package of nicotinamide is perfect for biochemical research involving NAD+ (Nicotinamide adenine dinucleotide), a crucial coenzyme in redox reactions within cells. Nicotinamide plays a vital role in maintaining skin health, DNA repair, and cellular energy production. It is frequently used in studies of metabolic pathways and cell signaling, especially in the context of vitamins and their role in health. Nicotinamide is also utilized in the development of therapeutic agents for treating skin disorders and certain neurological conditions.
Catalog No. 50-006-60833 Supplier Alkali Scientific Supplier No. COT001CP
Nicotinamide (Vitamin PP) is a key biochemical reagent used in a variety of applications related to cellular metabolism and enzymatic activity. This 100g package of nicotinamide is perfect for biochemical research involving NAD+ (Nicotinamide adenine dinucleotide), a crucial coenzyme in redox reactions within cells. Nicotinamide plays a vital role in maintaining skin health, DNA repair, and cellular energy production. It is frequently used in studies of metabolic pathways and cell signaling, especially in the context of vitamins and their role in health. Nicotinamide is also utilized in the development of therapeutic agents for treating skin disorders and certain neurological conditions.