Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nicotine Bi-L-(+)-tartrate Dihydrate 98.0+%, TCI America™
SDP

Catalog No. N008025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N008025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N008025G Supplier TCI America Supplier No. N008025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity laboratory grade chemical intended for research applications.

Chemical Identifiers

CAS 6-3-6019
Molecular Formula C18H30N2O14
Molecular Weight (g/mol) 498.44
MDL Number MFCD00150899
InChI Key LDMPZNTVIGIREC-HYRPJEHINA-N
Synonym nicotine ditartrate dihydrate, unii-r7m676m8yv, nicotine bitartrate usan, 2r,3r-2,3-dihydroxybutanedioic acid;3-2s-1-methylpyrrolidin-2-yl pyridine;dihydrate, nicotine bitartrate who-dd, nicotine acid tartrate dihydrate, s-3-1-methylpyrrolidin-2-yl pyridine bis 2r,3r-2,3-dihydroxysuccinate dihydrate, 2r,3r-2,3-dihydroxybutanedioic acid; 3-2s-1-methylpyrrolidin-2-yl pyridine; dihydrate, 3-2s-1-methyl-2-pyrrolidinyl pyridine, 2r,3r-2,3-dihydroxybutanedioate 1:2 , dihydrate
IUPAC Name bis((2R,3R)-2,3-dihydroxybutanedioic acid) 3-[(2S)-1-methylpyrrolidin-2-yl]pyridine dihydrate
SMILES O.O.O[C@H]([C@@H](O)C(O)=O)C(O)=O.O[C@H]([C@@H](O)C(O)=O)C(O)=O.CN1CCC[C@H]1C1=CN=CC=C1

Specifications

Color White-Yellow
Quantity 25 g
UN Number 1659
Formula Weight 462.41
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Nicotine Bi-L-(+)-tartrate Dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.