Learn More
Description
- Nicotine bitartrate dihydrate is naturally occurring isomer
- It is a Prototype nicotinic acetylcholine receptor agonist
Specifications
Specifications
| Melting Point | 93°C to 95°C (literature) |
| Color | White |
| Linear Formula | C10H14N2 . 2C4H6O6 . 2H2O |
| Quantity | 10 g |
| Solubility Information | Soluble in Water (50mg/mL, >10%). |
| Formula Weight | 498.44 |
| Percent Purity | 99.87% |
| Grade | Not Available |
| Physical Form | Powder |
| Chemical Name or Material | Nicotine bitartrate dihydrate |
Safety and Handling
| Recommended Storage | Store at Room Temperature(15-30°C). |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.