Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nicotine ditartrate dihydrate, 98%

Catalog No. p-172352
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC415660100 10 g
AC415660500 50 g
AC415661000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC415660100 Supplier Thermo Scientific Chemicals Supplier No. 415660100
Only null left
Add to Cart

CAS: 6019-06-3 | C18H30N2O14 | 498.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6-3-6019
Molecular Weight (g/mol) 498.44
MDL Number MFCD00150899
InChI Key LDMPZNTVIGIREC-HYRPJEHINA-N
Synonym (-)-Nicotine di-(+)-hydrogen tartrate dihydrate
IUPAC Name N-{3-[(2Z)-5-chloro-2,3-dihydro-1,3-benzoxazol-2-ylidene]-4-oxocyclohexa-1,5-dien-1-yl}-2-methylpropanamide
SMILES O.O.O[C@H]([C@@H](O)C(O)=O)C(O)=O.O[C@H]([C@@H](O)C(O)=O)C(O)=O.CN1CCC[C@H]1C1=CN=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 97.0°C to 100.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C18H30N2O14
Quantity 10 g
Solubility Information Solubility in water: 50g/L (20°C)
Formula Weight 498.44
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material Nicotine ditartrate dihydrate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic to aquatic life with long lasting effects.
Fatal in contact with skin.
Fatal if inhaled.
Precautionary Statement GHS P Statement
IF ON SKIN: Gently wash with plenty of soap and water.
Wear protective gloves/protective clothing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.