Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-Nicotine Solution, For drug analysis, 1.0 mg/mL in Methanol, MilliporeSigma™ Supelco™

Catalog No. 111017882 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-7882 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-7882 Supplier MilliporeSigma Supelco Supplier No. N55111ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Featured Industries: Agriculture, Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 54-11-5
Linear Formula C10H14N2
MDL Number MFCD00006369
Type Analytical Standard
Industry Type Drug Analysis
Quantity 1 mL
Formula Weight 162.23
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 1.0 mg/mL in methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.