Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nicotine Solution NMR Reference Standard, 2 mg/mL in DMSO-d6 (99.9 atom D), TMS 0.05, NMR tube size 5 mm, MilliporeSigma™ Supelco™

Catalog No. 111002892 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-2892 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-2892 Supplier MilliporeSigma Supelco Supplier No. 61203010X8IN
Add to Cart
Add to Cart

Specifications

Packaging Each tube contains 0.700 mL. Each Pk. contains 10 tubes.
Flash Point 87°C
Concentration 2 mg/mL in DMSO- d 6 (99.9 atom% D)
Linear Formula C10H14N2
Type NMR Reference Standard
Quantity 1 Ea.
Formula Weight 162.23
Grade NMR Reference Standard
Concentration or Composition (by Analyte or Components) TMS, 0.05%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.