Learn More
Specifications
Specifications
| CAS | 1504589 |
| Melting Point | 93°C to 95°C (literature) |
| Color | White |
| pH | 3.18 (10% aq. soln.) (literature) |
| Quantity | 1 g |
| Synonym | 4-pentynoic acid, propargylacetic acid, pent-4-yn-1-oic acid, 4-pentynoicacid, pent4-ynoic acid, zlchem 796, pubchem3491, pent-4-yn-oic acid, acmc-1az7d |
| Solubility Information | Soluble in water (50mg/mL - clear to slightly hazy, colorless to faint yellow solution) or alcohol |
| IUPAC Name | pent-4-ynoic acid |
| Formula Weight | 498.44 |
| Grade | Not Available |
| Show More |
Safety and Handling
| Recommended Storage | Store at Room Temperature (15 to 30°C) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.