Learn More
Cayman Chemical Nicotinic Acid, Purity: ≥98%, CAS: 59-67-6, 500 g
SDP
Catalog No. 502917602
Encompass_Preferred
An HCA2 antagonist (Ki = 113 nM); reduces lipolysis in primary human adipocytes (IC50 = 200 nM); reduces the invasion of AH109A-TC cancer cells at 20 and 40 µM; increases cutaneous vasodilation in normotensive mice in a dose-dependent manner
Catalog No. 50-291-7602 Supplier Cayman Chemical Supplier No. 37416500 G
An HCA2 antagonist (Ki = 113 nM); reduces lipolysis in primary human adipocytes (IC50 = 200 nM); reduces the invasion of AH109A-TC cancer cells at 20 and 40 µM; increases cutaneous vasodilation in normotensive mice in a dose-dependent manner