Learn More
Medchemexpress LLC Nicotinic acid mononucleotide | 321-02-8 | MFCD00070358 | 98.1% | 335.2 g·mol⁻1 | C11H14NO9P | 5 MG
SDP
Catalog No. 5000262768
Encompass_Preferred
Nicotinic acid mononucleotide (NaMN) is a nucleotide intermediate in the biosynthesis of NAD+. It is produced from nicotinic acid via phosphoribosyltransferase and serves as a substrate for adenylyltransferases involved in NAD+ metabolism. Supplied as a research-grade reagent for biochemical and enzymatic studies.
  • Intermediate in NAD+ biosynthesis and metabolism.
  • Substrate for adenylyltransferase and enzymatic assays.
  • High purity, typically ~98%.
  • Molecular weight 335.2 g·mol⁻1; formula C11H14NO9P.
  • Intended for research use only; not for human consumption.
Catalog No. 50-002-62768 Supplier Medchemexpress LLC Supplier No. HY1287005MG
Nicotinic acid mononucleotide (NaMN) is a nucleotide intermediate in the biosynthesis of NAD+. It is produced from nicotinic acid via phosphoribosyltransferase and serves as a substrate for adenylyltransferases involved in NAD+ metabolism. Supplied as a research-grade reagent for biochemical and enzymatic studies.
  • Intermediate in NAD+ biosynthesis and metabolism.
  • Substrate for adenylyltransferase and enzymatic assays.
  • High purity, typically ~98%.
  • Molecular weight 335.2 g·mol⁻1; formula C11H14NO9P.
  • Intended for research use only; not for human consumption.