Learn More
Medchemexpress LLC Nicotinic acid mononucleotide triethylamine | 00-00-0 | MFCD00070358 | 99.4% | 506.97 | C11H14NO9P.1.7C6H15N | 5 MG
SDP
Catalog No. 5000264053
Encompass_Preferred
Nicotinic acid mononucleotide triethylamine is the triethylamine salt of nicotinic acid mononucleotide, an intermediate in the biosynthesis of NAD+. It is supplied for research use in biochemical and metabolic studies and is offered in small milligram pack sizes with supporting analytical documentation.
  • High purity suitable for biochemical research.
  • Triethylamine salt form to aid handling and solubility.
  • Functionally relevant intermediate in NAD+ biosynthesis studies.
  • Available in small-scale quantities for laboratory workflows.
  • COA and SDS available for quality and safety information.
Catalog No. 50-002-64053 Supplier Medchemexpress LLC Supplier No. HY128700A5MG
Nicotinic acid mononucleotide triethylamine is the triethylamine salt of nicotinic acid mononucleotide, an intermediate in the biosynthesis of NAD+. It is supplied for research use in biochemical and metabolic studies and is offered in small milligram pack sizes with supporting analytical documentation.
  • High purity suitable for biochemical research.
  • Triethylamine salt form to aid handling and solubility.
  • Functionally relevant intermediate in NAD+ biosynthesis studies.
  • Available in small-scale quantities for laboratory workflows.
  • COA and SDS available for quality and safety information.