Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nicotinic acid mononucleotide triethylamine | 99.4% | 100 MG
SDP

Catalog No. 5000341029
Encompass_Preferred

Nicotinic acid mononucleotide triethylamine is a crucial intermediate formed from nicotinic acid (NA) during the biosynthesis of NAD+. It acts as a substrate for nicotinamide mononucleotide/nicotinic acid mononucleotide adenylyltransferase, playing a vital role in metabolic pathways.

  • Formed from nicotinic acid via nicotinic acid phosphoribosyltransferase
  • Substrate for nicotinamide mononucleotide/nicotinic acid mononucleotide adenylyltransferase
  • Molecular weight: 506.97
  • Formula: C11H14NO9P.1.7C6H15N
  • Appearance: Solid
  • Color: White to light yellow
  • Storage: 4°C, sealed, away from moisture; in solvent: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-41029 Supplier Medchemexpress LLC Supplier No. HY128700100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nicotinic acid mononucleotide triethylamine is a crucial intermediate formed from nicotinic acid (NA) during the biosynthesis of NAD+. It acts as a substrate for nicotinamide mononucleotide/nicotinic acid mononucleotide adenylyltransferase, playing a vital role in metabolic pathways.

  • Formed from nicotinic acid via nicotinic acid phosphoribosyltransferase
  • Substrate for nicotinamide mononucleotide/nicotinic acid mononucleotide adenylyltransferase
  • Molecular weight: 506.97
  • Formula: C11H14NO9P.1.7C6H15N
  • Appearance: Solid
  • Color: White to light yellow
  • Storage: 4°C, sealed, away from moisture; in solvent: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.