Learn More
Medchemexpress LLC Nicotinic acid mononucleotide triethylamine | 99.4% | 50 MG
SDP
Catalog No. 5000341920
Encompass_Preferred
Nicotinic acid mononucleotide triethylamine is formed from nicotinic acid (NA) via the nicotinic acid phosphoribosyltransferase in the biosynthesis of NAD+. It acts as a substrate for nicotinamide mononucleotide/nicotinic acid mononucleotide adenylyltransferase. For research use only.
  • Formed from nicotinic acid (NA)
  • Acts as a substrate for nicotinamide mononucleotide/nicotinic acid mononucleotide adenylyltransferase
  • Involved in the biosynthesis of NAD+
  • Endogenous metabolite
  • For research use only
Catalog No. 50-003-41920 Supplier Medchemexpress LLC Supplier No. HY12870050MG
Nicotinic acid mononucleotide triethylamine is formed from nicotinic acid (NA) via the nicotinic acid phosphoribosyltransferase in the biosynthesis of NAD+. It acts as a substrate for nicotinamide mononucleotide/nicotinic acid mononucleotide adenylyltransferase. For research use only.
  • Formed from nicotinic acid (NA)
  • Acts as a substrate for nicotinamide mononucleotide/nicotinic acid mononucleotide adenylyltransferase
  • Involved in the biosynthesis of NAD+
  • Endogenous metabolite
  • For research use only