Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nicotinic acid mononucleotide triethylamine | 99.89% | 50 MG
SDP

Catalog No. 5000339739
Encompass_Preferred

Nicotinic acid mononucleotide triethylamine is formed from nicotinic acid (NA) via the nicotinic acid phosphoribosyltransferase in the biosynthesis of NAD+. It acts as a substrate for nicotinamide mononucleotide/Nicotinic acid mononucleotide adenylyltransferase.

  • Solid, white to light yellow appearance.
  • Shipped at room temperature in continental US.
  • Store at 4°C, sealed, away from moisture.
  • For solutions, store at -80°C for 6 months or -20°C for 1 month (sealed, away from moisture).
  • Soluble in DMSO (100 mg/mL, 197.25 mM).
  • Soluble in H2O (100 mg/mL, 197.25 mM).
  • For research use only; not validated for medical applications.

Catalog No. 50-003-39739 Supplier Medchemexpress LLC Supplier No. HY128700A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nicotinic acid mononucleotide triethylamine is formed from nicotinic acid (NA) via the nicotinic acid phosphoribosyltransferase in the biosynthesis of NAD+. It acts as a substrate for nicotinamide mononucleotide/Nicotinic acid mononucleotide adenylyltransferase.

  • Solid, white to light yellow appearance.
  • Shipped at room temperature in continental US.
  • Store at 4°C, sealed, away from moisture.
  • For solutions, store at -80°C for 6 months or -20°C for 1 month (sealed, away from moisture).
  • Soluble in DMSO (100 mg/mL, 197.25 mM).
  • Soluble in H2O (100 mg/mL, 197.25 mM).
  • For research use only; not validated for medical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.